Skip to Content
Merck
All Photos(5)

Documents

R0250

Sigma-Aldrich

D-(+)-Raffinose pentahydrate

≥98.0% (HPLC)

Synonym(s):

O-α-D-Galactopyranosyl-(1→6)-α-D-glucopyranosyl β-D-fructofuranoside, Melitose, Melitriose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H32O16 · 5H2O
CAS Number:
Molecular Weight:
594.51
Beilstein:
99541
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

plant (Gossypium hirsutum L./Cotton)

Quality Level

Assay

≥98.0% (HPLC)

form

powder

technique(s)

HPLC: suitable

color

white

solubility

water: 50 mg/mL, clear, colorless

storage temp.

room temp

SMILES string

O.O.O.O.O.OC[C@H]1O[C@H](OC[C@H]2O[C@H](O[C@]3(CO)O[C@H](CO)[C@@H](O)[C@@H]3O)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C18H32O16.5H2O/c19-1-5-8(22)11(25)13(27)16(31-5)30-3-7-9(23)12(26)14(28)17(32-7)34-18(4-21)15(29)10(24)6(2-20)33-18;;;;;/h5-17,19-29H,1-4H2;5*1H2/t5-,6-,7-,8+,9-,10-,11+,12+,13-,14-,15+,16+,17-,18+;;;;;/m1...../s1

InChI key

BITMAWRCWSHCRW-PFQJHCPISA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Raffinose is a trisaccharide composed of galactose, fructose, and glucose. Raffinose is hydrolysed to D-galactose and sucrose by D-galactosidase.

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charlotte Holme Nielsen et al.
Developmental neuroscience, 40(5-6), 586-600 (2019-05-30)
Preterm birth interrupts intrauterine brain growth and maturation and may induce a delay in postnatal neurodevelopment. Such developmental delays can result from the reduced fetal age at birth, together with the clinical compli-cations of preterm birth (e.g., hypoxia, ischemia, and
Jonathan B Thacker et al.
Journal of the American Society for Mass Spectrometry, 30(3), 439-447 (2018-12-19)
The relationship between the electrospray ionization (ESI)-mass spectrometric (MS) response of an analyte and its concentration has been well studied for permanently charged and basic analytes in the positive ionization mode, but there has been a lack of research effort
A Lüdeking et al.
FEBS letters, 439(1-2), 180-184 (1998-12-16)
The gastrointestinal tract is exposed to environmental insult as a result of food intake or in pathological conditions such as diarrhoea, and is therefore protected by the mucus layer. As part of it, trefoil peptides (TFFs) are able to modify
Lise Dauban et al.
Journal of structural biology, 208(2), 152-164 (2019-08-27)
Spatial organisation of chromosomes is a determinant of genome stability and is required for proper mitotic segregation. However, visualization of individual chromatids in living cells and quantification of their geometry, remains technically challenging. Here, we used live cell imaging to
Benjamin M Vincent et al.
Cell reports, 25(10), 2742-2754 (2018-12-06)
The lack of disease-modifying treatments for neurodegenerative disease stems in part from our rudimentary understanding of disease mechanisms and the paucity of targets for therapeutic intervention. Here we used an integrated discovery paradigm to identify a new therapeutic target for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service