Skip to Content
Merck
All Photos(1)

Key Documents

95068

Supelco

D-(+)-Raffinose pentahydrate

analytical standard

Synonym(s):

O-α-D-Galactopyranosyl-(1→6)-α-D-glucopyranosyl β-D-fructofuranoside, Melitose, Melitriose

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$136.00

$136.00


Available to ship on07 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 MG
$136.00

About This Item

Empirical Formula (Hill Notation):
C18H32O16 · 5H2O
CAS Number:
Molecular Weight:
594.51
Beilstein:
99541
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.24

$136.00


Available to ship on07 April 2025Details


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

optical activity

[α]/D 105±2°, c = 10 in H2O

shelf life

limited shelf life, expiry date on the label

analyte chemical class(es)

oligosaccharides

impurities

13-17% water

application(s)

food and beverages

format

neat

SMILES string

O.O.O.O.O.OC[C@H]1O[C@H](OC[C@H]2O[C@H](O[C@]3(CO)O[C@H](CO)[C@@H](O)[C@@H]3O)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C18H32O16.5H2O/c19-1-5-8(22)11(25)13(27)16(31-5)30-3-7-9(23)12(26)14(28)17(32-7)34-18(4-21)15(29)10(24)6(2-20)33-18;;;;;/h5-17,19-29H,1-4H2;5*1H2/t5-,6-,7-,8+,9-,10-,11+,12+,13-,14-,15+,16+,17-,18+;;;;;/m1...../s1

InChI key

BITMAWRCWSHCRW-PFQJHCPISA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Raffinose pentahydrate is a crystalline form of pharmaceutical sugar, raffinose which may act as ′scavengers′ or ′reservoirs′ for water with the pharmaceutical formulations.[1]

Application

D-(+)-Raffinose pentahydrate may be used as a standard for the determination of free sugars of plants for separating the trimethylsilyl ethers by gas chromatography method.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Gas chromatography of carbohydrates the quantitative determination of the free sugars of plants as their trimethylsilyl ethers.
Davison PK and Young R
Journal of Chromatography A, 41(8), 12-21 (1969)
Water vapour sorption by pharmaceutical sugars.
Hancock BC and Shamblin SL
Pharmaceutical Science and Technology Today, 1(8), 345-351 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service