Skip to Content
Merck
All Photos(2)

Documents

P8582

Sigma-Aldrich

Podophyllin from Podophyllum emodii

powder

Synonym(s):

Podophyllum resin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.83

form

powder

color

brown

storage temp.

2-8°C

InChI

1S/2C22H24O9.2C22H22O8/c2*1-27-16-4-10(5-17(28-2)21(16)29-3)18-11-6-14-15(31-9-30-14)7-12(11)20(24)13(8-23)19(18)22(25)26;2*1-25-16-4-10(5-17(26-2)21(16)27-3)18-11-6-14-15(30-9-29-14)7-12(11)20(23)13-8-28-22(24)19(13)18/h2*4-7,13,18-20,23-24H,8-9H2,1-3H3,(H,25,26);2*4-7,13,18-20,23H,8-9H2,1-3H3

InChI key

KOUZYZCESNTXJO-UHFFFAOYSA-N

General description

Podophyllin is an alcoholic plant extract, obtained from the dried rhizome and roots of Podoplryllui emodi. It is a powder with colors varying from light brown to greenish-yellow and exposure to heat or light turns it darker.

Biochem/physiol Actions

Podophyllin is used to treat condylomata acuminate, a type of venereal warts. It exhibit several biological functions and is mainly used in the treatment of constipation, hepatic disorder and rheumatic arthritis.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Eye Dam. 1 - Repr. 1B - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Condylomata acuminata: two hundred cases treated with podophyllin
Culp OS and Kaplan IW
Annals of Surgery, 120(2), 251-251 (1944)
Simultaneous determination of podophyllotoxin, quercetin and kaempferol in podophyllin by liquid chromatography tandem mass spectrometry
Lin M C, et al.
Journal of food and drug analysis, 16(6), 251-251 (2008)
Podophyllin and its use in the treatment of condylomata acuminata
Sundharam J A, et al.
Indian Journal of Dermatology, Venereology and Leprology, 56(1), 10-10 (1990)
Condylomata acuminata in infants and young children. Topical podophyllin an effective therapy.
Khalifa E Sharquie et al.
Saudi medical journal, 26(3), 502-503 (2005-04-05)
L CostaPinto et al.
Lupus, 21(3), 332-334 (2011-10-04)
Females with systemic lupus erythematosus (SLE) have higher prevalence of human papillomavirus (HPV) infection, which can lead to the development of warts. Herein we report the first case of giant disseminated condylomatosis (GDC) in a SLE female on mycophenolate mofetil

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service