Skip to Content
Merck
All Photos(1)

Key Documents

371708

Sigma-Aldrich

Gβγ Modulator II, Gallein

A cell-permeable xanthene compound that binds to Gβγ with high affinity (Kd ~400 nM derived from a surface plasma resonance binding study using Gβ1γ2) and blocks Gβγ-dependent cellular activities.

Synonym(s):

Gβγ Modulator II, Gallein, 4,5-Dihydroxyfluorescein, dihydrate, 2-(3,4,5-Trihydroxy-6-oxoxanthen-9-yl)-benzoic acid, dihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$233.00

$233.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$233.00

About This Item

Empirical Formula (Hill Notation):
C20H12O7 · 2H2O
CAS Number:
Molecular Weight:
400.34
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.77

$233.00


Estimated to ship on30 May 2025


Request a Bulk Order

Quality Level

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

burgundy

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

O1C3(c4c(c(c(cc4)O)O)Oc5c3ccc(c5O)O)c2c(cccc2)C1=O

InChI

1S/C20H12O7/c21-13-7-5-11-17(15(13)23)26-18-12(6-8-14(22)16(18)24)20(11)10-4-2-1-3-9(10)19(25)27-20/h1-8,21-24H

InChI key

PHLYOKFVXIVOJC-UHFFFAOYSA-N

General description

A cell-permeable xanthene compound that binds to Gβγ with high affinity (Kd ~400 nM derived from a surface plasma resonance binding study using Gβ1γ2) and blocks Gβγ-dependent cellular activities. Reported to exhibit in vivo prophylactic efficacy against carrageenan-induced neutrophil recruitment and tissue swelling in a murine paw edema model via either intraperitoneal or oral administration.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Lehmann, D.M., et al. 2008. Mol. Pharmacol.73, 410.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service