Skip to Content
Merck
All Photos(1)

Key Documents

P1997

Sigma-Aldrich

Anti-phospho-Dynamin (pSer774) antibody produced in sheep

affinity isolated antibody, aqueous glycerol suspension

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
$806.00

$806.00


Check Cart for Availability


Select a Size

Change View
1 VIAL
$806.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$806.00


Check Cart for Availability

biological source

sheep

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

aqueous glycerol suspension

usage

sufficient for 10 blots

species reactivity

rat, mouse, human

technique(s)

immunohistochemistry (frozen sections): suitable
indirect immunofluorescence: suitable
western blot: 1:1000 using using rat brain synaptosomal lysate.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Immunogen

synthetic phosphopeptide corresponding to amino acid residues surrounding Ser774 of dynamin.

Physical form

Solution in 10 mM HEPES, pH 7.5, 150 mM NaCl, 100 μg/mL BSA, and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service