Skip to Content
Merck
All Photos(1)

Key Documents

AB1892

Sigma-Aldrich

Anti-C-X-X-X-C Chemokine Receptor 1 Antibody, NT

Chemicon®, from rabbit

Synonym(s):

CX3CR1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$542.00

$542.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$542.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$542.00


Estimated to ship on30 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, human, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CX3CR1(1524)

General description

CX3CR1 is one of the chemokine receptors that are required as coreceptors for HIV infection. The genes encoding human, mouse, and rat CX3CR1 were cloned and designated V28 and CMKBRL1, CX3CR1, and RBS11, respectively (1-4). The seven transmembrane protein was recently identified as the receptor for a novel transmembrane molecule, fractalkine, and renamed CX3CR1 (5). Recently, CX3CR1 was found to serve as a coreceptor for HIV-1 and HIV-2 envelope fusion and virus infection, which can be inhibited by facktokine (6). CX3CR1 mediates leukocyte migration and adhesion.

Specificity

Specific for human C-X-X-X-C Chemokine Receptor 1 (CX3CR1) at the Extracellular loop. The sequence is identical to that of rat and differs from that of mouse by only one amino acid.

Immunogen

Epitope: N-terminus
Synthetic peptide from the N-terminal of human CX3CR1.

Application

Detect C-X-X-X-C Chemokine Receptor 1 using this Anti-C-X-X-X-C Chemokine Receptor 1 Antibody, N-terminus validated for use in IH & WB.
Research Category
Inflammation & Immunology
Research Sub Category
Cytokines & Cytokine Receptors
Western blot: 1:500 - 1:2000 (Human spleen tissue lysate can be used as positive control and an approximately 50 kDa band can be detected)

Immunohistochemistry in frozen sections

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
Purified immunoglobulin. 100 μg purified IgG in 100 μL of PBS containing 0.2% sodium azide.

Storage and Stability

Store at 2-8°C for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xilei Li et al.
Frontiers in neuroscience, 15, 620525-620525 (2021-04-13)
Previous studies have proved that peripheral nerve injury is involved in the pathogenesis of neuropathic pain (NP). The peripheral nerve injury primes spinal M1 microglia phenotype and produces pro-inflammatory cytokines, which are responsible for neurotoxic and neuronal hyper-excitable outcomes. Spinal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service