Skip to Content
Merck
All Photos(1)

Key Documents

P0869

Sigma-Aldrich

Monoclonal Anti-Peroxisome Proliferator Activated Receptor α antibody produced in mouse

~1 mg/mL, clone 3B6/PPAR, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-PPARα

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$566.00

$566.00


Estimated to ship on02 June 2025



Select a Size

Change View
100 μG
$566.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$566.00


Estimated to ship on02 June 2025


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3B6/PPAR, monoclonal

form

buffered aqueous solution

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

western blot: 2-5 μg/mL using human, mouse and rat

isotype

IgG2b

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PPARA(5465)
mouse ... Ppara(19013)
rat ... Ppara(25747)

General description

Peroxisome proliferator-activated receptors (PPARs) are a group of closely related nuclear receptors. Three PPAR isotypes have been identified: α,β (also called NUC1) and γ. PPARa mRNA is expressed most in brown adipose tissue, heart, liver and kidney in rats. The gene encoding PPARα is localized on human chromosome 22q12-q13.1 and rat chromosome 7q34.

Immunogen

purified recombinant PPARα protein.

Biochem/physiol Actions

Peroxisome proliferator-activated receptors (PPARs) may be involved in chronic diseases such as diabetes, obesity, atherosclerosis and cancer. PPARα has a protective function in fatty acids overload associated tubular injury. The enzyme also has a role in fatty acid oxidation and metabolism.

Physical form

Solution in phosphate buffered saline containing 1 mg/ml bovine serum albumin and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria L R Hughes et al.
The Journal of biological chemistry, 290(22), 13895-13906 (2015-04-08)
Nuclear hormone receptors (NHRs) regulate the expression of proteins that control aspects of reproduction, development and metabolism, and are major therapeutic targets. However, NHRs are ubiquitous and participate in multiple physiological processes. Drugs that act at NHRs are therefore commonly
Peroxisome Proliferator-Activated Receptors: Nuclear Control of Metabolism*
Be?atrice Desvergne and Walter Wahli
Endocrine Reviews, 20(5), 649?688-649?688 (1999)
Fenofibrate, a PPAR? agonist, protect proximal tubular cells from albumin-bound fatty acids induced apoptosis via the activation of NF-kB
Nan Zuo
International Journal of Clinical and Experimental Pathology, 8(9), 10653?10661-10653?10661 (2015)
C W Park et al.
Kidney international, 69(9), 1511-1517 (2006-05-05)
Peroxisome proliferator-activated receptor alpha (PPARalpha) is a member of the ligand-activated nuclear receptor superfamily, and plays an important role in lipid metabolism and glucose homeostasis. The purpose of this study is to determine whether the activation of PPARalpha by fenofbrate
PPARs: molecular targets in the pharmacogenomics era
Prague Medical Report, 223-236 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service