Skip to Content
Merck
All Photos(1)

Key Documents

P0059

Sigma-Aldrich

Pro-carboxypeptidase B human

≥2.0 units/mg protein

Synonym(s):

Thrombin Activatable Fibrinolysis Inhibitor (TAFI)

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$959.00

$959.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
50 μG
$959.00

About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

$959.00


Estimated to ship on30 May 2025


Request a Bulk Order

Quality Level

specific activity

≥2.0 units/mg protein

mol wt

60 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CPB1(1360)

Biochem/physiol Actions

TAFI is supplied in its 60 kDa zymogen form. The N-terminal of TAFI contains four N-linked glycans and a plasminogen recognition sequence. Hydrolysis of the activation peptide by thrombin releases a 36-kDa active proteolytic domain homologous to tissue carboxypeptidase B. Complexation with thrombomodulin increases thrombin activation of TAFI 1250 fold. TAFI exerts it′s inhibition of fibrinolytic activity by binding to plasminogen and by cleaving C-terminal lysine residues of fibrin resulting in the reduction of plasmin and tPA binding sites on fibrin.[1] Experiments in TAFI-deficient mice point to a regulatory role on complement C5a.[2]

Unit Definition

One unit will hydrolyze 1 micromole of Hippuryl-L-Arginine per minute at pH 7.4 at 25 °C.

Physical form

Solution containing 20 mM Hepes and 150 mM Sodium chloride. pH 7.4

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toshihiko Nishimura et al.
Blood, 109(5), 1992-1997 (2006-11-16)
Plasma procarboxypeptidase B (proCPB) is activated by the endothelial thrombin-thrombomodulin [corrected] complex. Activated proCPB [corrected] (CPB) functions as a fibrinolysis inhibitor, but it may play a broader role by inactivating inflammatory mediators. To test this hypothesis, C5a-induced alveolitis was studied
B N Bouma et al.
Journal of thrombosis and haemostasis : JTH, 1(7), 1566-1574 (2003-07-23)
Recently, a new inhibitor of fibrinolysis was described, which downregulated fibrinolysis after it was activated by thrombin, and was therefore named TAFI (thrombin-activatable fibrinolysis inhibitor; EC 3.4.17.20). TAFI turned out to be identical to the previously described proteins, procarboxypeptidase U

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service