Skip to Content
Merck
All Photos(1)

Documents

P0039

Sigma-Aldrich

PHPS1 sodium salt hydrate

≥98% (HPLC)

Synonym(s):

4-[2-[1,5-dihydro-3-(4-nitrophenyl)-5-oxo-1-phenyl-4H-pyrazol-4-ylidene]hydrazinyl]-benzenesulfonic acid sodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H15N5O6S · xNa+ · yH2O
CAS Number:
Molecular Weight:
465.44 (anhydrous free acid basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

color

red

solubility

DMSO: 10 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O=C1/C(C(C2=CC=C([N+]([O-])=O)C=C2)=NN1C3=CC=CC=C3)=N/NC4=CC=C(S(=O)([O-])=O)C=C4.[Na+].O

InChI

1S/C21H15N5O6S.Na.H2O/c27-21-20(23-22-15-8-12-18(13-9-15)33(30,31)32)19(14-6-10-17(11-7-14)26(28)29)24-25(21)16-4-2-1-3-5-16;;/h1-13,22H,(H,30,31,32);;1H2/q;+1;/p-1/b23-20+;;

InChI key

KLXUVMQUEDKCKG-JBKNFAMTSA-M

Application

Phenylhydrazono pyrazolone sulfonate 1 (PHPS1) sodium salt hydrate has been used as a protein-tyrosine phosphatase (shp2) inhibitor to study its effects on bisphenol A (BPA) and nonyl phenol (NP)-induced cell proliferation of MCF7 cell. It has also been used as a shp2 inhibitor to study its pro-atherosclerotic or atheroprotective effect in vivo and in vitro.

Biochem/physiol Actions

Phenylhydrazono pyrazolone sulfonate 1 (PHPS1) represses E2-induced gene transcription and rapid DNA synthesis. It also blocks mitogen-activated protein kinase (ERK1/2) activation and reduces the cigarette smoke induced-inflammatory response in mouse lungs.
Shp-2 is a Src Homology-2 (SH2) containing phosphatase that regulates growth factor signalling. The enzyme is expressed ubiquitously in mammalian tissues and plays essential roles in development. It is also implicated in various pathological states. PHPS1 is a selective inhibitor of Shp-2 and blocks Shp-2 dependent downstream signalling.

Features and Benefits

This compound is featured on the Phosphoprotein Phosphatases (Tyrosine) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunmei Fan et al.
Cell reports, 39(4), 110762-110762 (2022-04-28)
Tendon maturation lays the foundation for postnatal tendon development, its proper mechanical function, and regeneration, but the critical cell populations and the entangled mechanisms remain poorly understood. Here, by integrating the structural, mechanical, and molecular properties, we show that post-natal
SHP2 inhibitor PHPS1 protects against atherosclerosis by inhibiting smooth muscle cell proliferation
Chen J, et al.
BMC Cardiovascular Disorders, 18(1), 72-72 (2018)
Increased Cell Proliferation by Various Doses of Xenoestrogen in MFC-7 Cell Through Tyrosine Phosphatase Shp2
Liu W, et al.
American Journal of Bioscience and Bioengineering, 3(6), 178-182 (2015)
Heng Zhou et al.
Journal of molecular medicine (Berlin, Germany), 98(5), 691-705 (2020-04-14)
Cardiac hypertrophy is a complex pathological process, and the molecular mechanisms underlying hypertrophic remodeling have not been clearly elucidated. Leukocyte immunoglobulin-like receptor B4 (lilrb4) is an inhibitory transmembrane protein that is necessary for the regulation of various cellular signaling pathways.
Tyrosine phosphatase Shp2 mediates the estrogen biological action in breast cancer via interaction with the estrogen extranuclear receptor.
Li J, Kang Y, Wei L, et al.
PLoS ONE, 9(7), e102847-e102847 (2014)

Articles

Protein tyrosine phosphatases' catalytic mechanism involves transient phosphorylation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service