Merck
  • A Cd9+Cd271+ stem/progenitor population and the SHP2 pathway contribute to neonatal-to-adult switching that regulates tendon maturation.
A Cd9+Cd271+ stem/progenitor population and the SHP2 pathway contribute to neonatal-to-adult switching that regulates tendon maturation.
Cell reports (2022-04-28)
Chunmei Fan, Yanyan Zhao, Yangwu Chen, Tian Qin, Junxin Lin, Shan Han, Ruojin Yan, Tingyun Lei, Yuanhao Xie, Tingzhang Wang, Shen Gu, Hongwei Ouyang, Weiliang Shen, Zi Yin, Xiao Chen
ABSTRACT
Tendon maturation lays the foundation for postnatal tendon development, its proper mechanical function, and regeneration, but the critical cell populations and the entangled mechanisms remain poorly understood. Here, by integrating the structural, mechanical, and molecular properties, we show that post-natal days 7-14 are the crucial transitional stage for mouse tendon maturation. We decode the cellular and molecular regulatory networks at the single-cell level. We find that a nerve growth factor (NGF)-secreting Cd9+Cd271+ tendon stem/progenitor cell population mainly prompts conversion from neonate to adult tendon. Through single-cell gene regulatory network analysis, in vitro inhibitor identification, and in vivo tendon-specific Shp2 deletion, we find that SHP2 signaling is a regulator for tendon maturation. Our research comprehensively reveals the dynamic cell population transition during tendon maturation, implementing insights into the critical roles of the maturation-related stem cell population and SHP2 signaling pathway during tendon differentiation and regeneration.
Sigma-Aldrich
PHPS1 sodium salt hydrate, ≥98% (HPLC)