O4500
1-Octanol
~99%, liquid
Synonym(s):
Alcohol C8, Capryl alcohol, Octyl alcohol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3(CH2)7OH
CAS Number:
Molecular Weight:
130.23
Beilstein:
1697461
EC Number:
MDL number:
UNSPSC Code:
12190000
Recommended Products
vapor density
4.5 (vs air)
vapor pressure
0.14 mmHg ( 25 °C)
Assay
~99%
form
liquid
autoignition temp.
523 °F
refractive index
n20/D 1.429 (lit.)
bp
196 °C (lit.)
mp
−15 °C (lit.)
density
0.827 g/mL at 25 °C (lit.)
SMILES string
CCCCCCCCO
InChI
1S/C8H18O/c1-2-3-4-5-6-7-8-9/h9H,2-8H2,1H3
InChI key
KBPLFHHGFOOTCA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Blocks T-type Ca2+ channels
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 3 - Eye Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
187.7 °F
Flash Point(C)
86.5 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Erich M Staudacher et al.
Journal of neuroscience methods, 180(2), 208-223 (2009-05-26)
A central problem facing studies of neural encoding in sensory systems is how to accurately quantify the extent of spatial and temporal responses. In this study, we take advantage of the relatively simple and stereotypic neural architecture found in invertebrates.
J Franco et al.
Acta biomaterialia, 6(1), 218-228 (2009-07-01)
The development of materials to support bone regeneration requires flexible fabrication technologies able to tailor chemistry and architecture for specific applications. In this work we describe the preparation of ceramic-based inks for robotic-assisted deposition (robocasting) using Pluronic F-127 solutions. This
Elian-Simplice Yaganza et al.
Applied and environmental microbiology, 75(5), 1465-1469 (2008-12-31)
Twenty-one salts were tested for their effects on the growth of Pectobacterium carotovorum subsp. carotovorum and Pectobacterium atrosepticum. In liquid medium, 11 salts (0.2 M) exhibited strong inhibition of bacterial growth. The inhibitory action of salts relates to the water-ionizing
Irene Sargiannidou et al.
Neurobiology of disease, 30(2), 221-233 (2008-03-21)
Murine oligodendrocytes express the gap junction (GJ) proteins connexin32 (Cx32), Cx47, and Cx29. CNS phenotypes in patients with X-linked Charcot-Marie-Tooth disease may be caused by dominant effects of Cx32 mutations on other connexins. Here we examined the expression of Cx31.3
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service