Skip to Content
Merck
All Photos(1)

Key Documents

N7395

Millipore

Immobilon®-NC HATF membranes

diam. 132 mm

Sign Into View Organizational & Contract Pricing

Select a Size

50 EA
$547.00

$547.00


Check Cart for Availability


Select a Size

Change View
50 EA
$547.00

About This Item

UNSPSC Code:
41105339
PubChem Substance ID:
NACRES:
NB.22

$547.00


Check Cart for Availability

form

solid

manufacturer/tradename

Immobilon®

diam.

132 mm

pore size

0.45 μm

Looking for similar products? Visit Product Comparison Guide

General description

Nitrocellulose with a small amount of cellulose acetate for improved handling. Triton-free. For colony hybridization, plaque lifts, colony growth directly on membrane.

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chao Zhang et al.
Cell cycle (Georgetown, Tex.), 14(13), 2100-2108 (2015-05-06)
Osteosarcoma is the most common primary malignant bone tumor and affects a significant portion of pediatric oncology patients. Although surgery and adjuvant chemotherapy confer significant survival benefits, many patients go on to develop metastatic disease, particularly to the lungs, secondary
Hsin-Chieh Lan et al.
Toxicological sciences : an official journal of the Society of Toxicology, 148(1), 26-34 (2015-07-26)
Certain commonly used compounds that interfere with the functions of the endocrine system are classified as endocrine-disrupting chemicals (EDCs). Bisphenol A (BPA) is an EDC that is widely used in food containers. BPA levels in human sera are commonly observed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service