Skip to Content
Merck
All Photos(1)

Documents

N1385

Sigma-Aldrich

Naringinase from Penicillium decumbens

≥300 units/g solid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352204

specific activity

≥300 units/g solid

secondary activity

≥10 units/g solid β-glucosidase

storage temp.

−20°C

Unit Definition

One unit will liberate 1.0 μmole of reducing sugar (measured as glucose) from naringin per min at pH 4.0 at 40 °C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hélder Vila-Real et al.
Enzyme research, 2011, 692618-692618 (2011-09-24)
The production of flavonoid glycosides by removing rhamnose from rutinosides can be accomplished through enzymatic catalysis. Naringinase is an enzyme complex, expressing both α-L-rhamnosidase and β-D-glucosidase activities, with application in glycosides hydrolysis. To produce monoglycosylated flavonoids with naringinase, the expression

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service