Skip to Content
Merck
All Photos(1)

Key Documents

487507

Sigma-Aldrich

p-Nitrophenyl-β-D-glucopyranoside

≥99% (HPLC), solid, β-glucosidase substrate, Calbiochem®

Synonym(s):

p-Nitrophenyl-β-D-glucopyranoside, p-Nitrophenyl-β-glucoside, 1-O- p-Nitrophenyl-D-glucose, p-Nitrophenyl-β-glucoside, 1-O-p-Nitrophenyl-D-glucose

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$98.20

$98.20


Estimated to ship on30 March 2025


Request a Bulk Order

Select a Size

Change View
1 G
$98.20

About This Item

Empirical Formula (Hill Notation):
C12H15NO8
CAS Number:
Molecular Weight:
301.25
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$98.20


Estimated to ship on30 March 2025


Request a Bulk Order

Product Name

p-Nitrophenyl-β-D-glucopyranoside, Chromogenic substrate for β-glucosidase.

Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

water: soluble

shipped in

ambient

storage temp.

10-30°C

SMILES string

[N+](=O)([O-])c1ccc(cc1)O[C@@H]2O[C@@H]([C@H]([C@@H]([C@H]2O)O)O)CO

InChI

1S/C12H15NO8/c14-5-8-9(15)10(16)11(17)12(21-8)20-7-3-1-6(2-4-7)13(18)19/h1-4,8-12,14-17H,5H2/t8-,9-,10+,11-,12-/m1/s1

InChI key

IFBHRQDFSNCLOZ-RMPHRYRLSA-N

General description

Chromogenic substrate for β-glucosidase.

Warning

Toxicity: Standard Handling (A)

Other Notes

Lin, J., et al. 1999. Biotechnol. Appl. Biochem.30, 81.
Riou, C., et al. 1998. Appl. Environ. Microbiol. 64, 3607.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Riou et al.
Applied and environmental microbiology, 64(10), 3607-3614 (1998-10-06)
Aspergillus oryzae was found to secrete two distinct beta-glucosidases when it was grown in liquid culture on various substrates. The major form had a molecular mass of 130 kDa and was highly inhibited by glucose. The minor form, which was
J Lin et al.
Biotechnology and applied biochemistry, 30(1), 81-87 (1999-09-01)
The beta-D-glucosidase produced by Thermomyces lanuginosus-SSBP was purified to apparent homogeneity. The purified enzyme consisted of two identical subunits with a native molecular mass of 200 kDa. The purified beta-D-glucosidase only hydrolysed the glucoside substrates containing a terminal, non-reducing beta-D-glucose

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service