MTOXRTSUP
RPTEC Tox Supplement
sterile-filtered, suitable for cell culture, add to 500 ML of basal medium
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
NACRES:
NA.81
Recommended Products
Quality Level
sterility
sterile-filtered
form
liquid
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−20°C
General description
Sigma has developed a variety of specialized media which are intended to provide optimal performance when utilized with specific cell lines. This product consists of 6 mL of RPTEC Tox media supplement which is added to basal media and other components listed below to produce 500 mL of media needed for optimal culture conditions of Sigma′s Human Renal Proximal Tubule Epithelial Cells (RPTEC) when used for Kidney Toxicology studies. The final RPTEC Tox media was developed specifically for use with Sigma′s human RPTEC control SA7K Clone cells and various knockouts (KO).
Preparation Note
Prepare Tox Medium by supplementing 500 mL of MEMα (M4526) with 6.25 mL of L-glutamine (G7513), 6 mL RPTEC Tox Supplement (MTOXRTSUP) and 0.3 mL of Gentamicin solution (G1397) and 0.03 mL of Amphotericin B solution (A2942). Tox Medium can be stored at 2–8 °C for up to 1 month. This medium is formulated for use with a 5% CO2 in air atmosphere.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Marianne K Vormann et al.
The AAPS journal, 20(5), 90-90 (2018-08-16)
Proximal tubules in the kidney play a crucial role in reabsorbing and eliminating substrates from the body into the urine, leading to high local concentrations of xenobiotics. This makes the proximal tubule a major target for drug toxicity that needs
Articles
AIM Biotech's modular platform enables simple to complex 3D cell culture for various research needs.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service