Skip to Content
Merck
All Photos(1)

Key Documents

MSG4000

Sigma-Aldrich

DC Mass Spect Gold® Serum

Synonym(s):

Gold Serum

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$1,020.00
300 ML
$1,960.00
1 L
$3,150.00

$1,020.00


Check Cart for Availability


Select a Size

Change View
100 ML
$1,020.00
300 ML
$1,960.00
1 L
$3,150.00

About This Item

UNSPSC Code:
12352204
NACRES:
NA.71
form:
liquid
shipped in:
dry ice

$1,020.00


Check Cart for Availability

grade

Proteomics Grade

form

liquid

shipped in

dry ice

storage temp.

−20°C

General description

DC Mass Spect Gold is processed from human plasma through a validated process that reduces the Hormone and Steroid levels, please see values below. This validated process maintains the inherent characteristics of the human plasma resulting in a pool that is consistent with patient samples. The human plasma is collected from donors at FDA licensed facilities. Each donor is screened for donor eligibility according to FDA regulations. Each unit is tested and found negative for HIV1/2-ab, HBsAg, HCV-ab, HIV-1 (NAT), HCV (NAT) and RPR. The plasmais inspected for conformance to raw material specifications. The plasma is processed according to a batch record. The finished product is analyzed and inspected according to quality control procedures for conformance to specifications. Records from donor unit(s) to final product are maintained according to established procedures. These records include donor records, raw material inspection records, batch records, testing records (baseline through final product), labeling records, final product inspection records and batch release records.

Application

Zero base or in preparation of controls, calibrators and standards for Immunoassays, GCMS and LCMS applications and Hormone and Steroid mass spectrometry application

Other Notes

Common Uses: Base matrix for hormone and steroid calibrators, controls, standards and linearity products.

Legal Information

Mass Spect Gold is a registered trademark of Golden West Biologicals, Inc.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service