Sign In to View Organizational & Contract Pricing.
Select a Size
100 ML
$1,060.00
$1,060.00
0 left in stock
About This Item
NACRES:
NA.71
UNSPSC Code:
12352204
Form:
liquid
Shipped in:
dry ice
Skip To
grade
Proteomics Grade
form
liquid
shipped in
dry ice
storage temp.
−20°C
1 of 4
This Item | |||
|---|---|---|---|
| form liquid | form liquid | form liquid | form liquid |
| shipped in dry ice | shipped in dry ice | shipped in dry ice | shipped in dry ice |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| grade Proteomics Grade | grade Proteomics Grade | grade Proteomics Grade | grade Proteomics Grade |
General description
VD-DC Mass Spect Gold® is human plasma that is processed according to a validated process, which removes the Vitamin D2 and Vitamin D3 to levels of < 1 ng/ml. The process maintains the inherent characteristics of the human plasma resulting in a Vitamin D product that is consistent with patient samples. The human plasma is collected from donors at FDA licensed facilities. Each donor is screened for donor eligibility according to FDA regulations. Each unit is tested and found negative for HIV1/2-ab, HBsAg, HCV-ab, HIV-1 (NAT), HCV (NAT) and RPR. The plasma is inspected for conformance to raw material specifications. The plasma is processed according to a batch record. The finished product is analyzed and inspected according to quality control procedures for conformance to specifications. Records from donor unit(s) to final product are maintained according to established procedures. These records include donor records, raw material inspection records, batch records, testing records (baseline through final product), labeling records, final product inspection records and batch release records.
Application
VD-DC Mass Spect Gold® Serum is used as a zero base or in preparation of controls, calibrators and standards for Immunoassays, GCMS and LCMS applications and Vitamin D mass spectrometry applications.
Common Uses: Base matrix for Vitamin D calibrators, controls, standards and linearity products.
Common Uses: Base matrix for Vitamin D calibrators, controls, standards and linearity products.
Other Notes
Each Unit of plasma used in the preparation of this material was tested and found negative or non-reactive for HIV 1/2, HBsAg, HCV, HIV-1 (NAT), HCV (NAT) and RPR by FDA approved methods. Products are intended for research or manufacturing of non-injectable products only. This material was tested as stated above in the testing section. Nevertheless, product of human origin should be considered potentially infectious and handled according to universal precautions. The safe and responsible handling of the product listed above is the responsibility of the customer. We accept no liability for injury or illness resulting from the use and/or handling of the product. The customer assumes responsibility for the proper disposal of all materials in accordance with Federal and State Regulations.
Legal Information
Mass Spect Gold is a registered trademark of Golden West Biologicals, Inc.
Disclaimer
In-vitro use only. Products are intended for research or manufacturing of non-injectible products only.
Storage Class
10 - Combustible liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service


