Skip to Content
Merck
All Photos(1)

Key Documents

MAK533

Sigma-Aldrich

Arginase Assay Kit

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$728.00

$728.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
1 KIT
$728.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

$728.00


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

application(s)

pharmaceutical

detection method

colorimetric

relevant disease(s)

gastrointestinal diseases; neurological disorders

shipped in

wet ice

storage temp.

−20°C

General description

Arginase (L-arginine ureohydrolase) is present in mammals and plants. In humans, arginase is expressed predominantly in the liver, and to lesser degrees in breast, kidney, testes, salivary glands, epidermis, and erythrocytes. Arginase catalyzes the conversion of arginine to ornithine and urea, completing the last step in the urea cycle. Arginase activity is a key diagnostic indicator. Increased levels of arginase activity in blood have been associated with liver damage. Hyperargininemia due to arginase deficiency is an inherited autosomal recessive disease.

Features and Benefits

  • Detection limit: 0.3 U/L
  • Compatible with high-throughput handling systems.

Suitability

The kit is suitable for the determination of arginase activity in enzyme preparations, serum, plasma, tissue culture etc., as well as for studying the effects of drugs on arginase activity.

Principle

The Arginase Assay Kit provides a sensitive and convenient method for arginase activity determination. The method utilizes a chromogen that forms a colored complex specifically with urea produced in the arginase reaction. The intensity of the color is directly proportional to the arginase activity in the sample.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Repr. 1B - Skin Corr. 1A

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service