Skip to Content
Merck
All Photos(1)

Key Documents

MAK521

Sigma-Aldrich

Fumarase Assay Kit

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$989.00

$989.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
$989.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

$989.00


Check Cart for Availability

Request a Bulk Order

Quality Level

detection method

colorimetric

relevant disease(s)

neurological disorders; neonatal diseases

shipped in

wet ice

storage temp.

−20°C

General description

Fumarase (or fumarate hydratase) is an enzyme that catalyzes the reversible hydration/dehydration reaction of fumarate to malate. Fumarase exists in two isoforms: a cytosolic and mitochondrial form. In the citric acid cycle, it facilitates a transition step in the production of energy in the form of NADH. Fumarase deficiency in humans results in early brain development problems and is characterized by poor feeding, hypotonia, failure to thrive, etc.

Features and Benefits

  • Linear detection range: 0.4 to 70 U/L
  • Compatible with high-throughput handling systems.

Suitability

The kit is suitable for fumarase activity determination in biological samples (e.g. plasma, serum, erythrocytes, tissue and culture media.)

Principle

The Fumarase Assay Kit is a non-radioactive, colorimetric assay based on the reduction of the tetrazolium salt MTT in a NADH-coupled enzymatic reaction to a reduced form of MTT which exhibits an absorption maximum at 565 nm. The increase in absorbance at 565 nm is proportional to the enzyme activity.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service