Skip to Content
Merck
All Photos(1)

Key Documents

MAK494

Sigma-Aldrich

Creatine Assay Kit

sufficient for 100 colorimetric or fluorometric tests

Synonym(s):

Creatine Quantification Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$731.00

$731.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 KIT
$731.00

About This Item

UNSPSC Code:
12352204
NACRES:
NA.84

$731.00


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

application(s)

food and beverages

detection method

colorimetric
fluorometric

relevant disease(s)

neurological disorders; musculoskeletal disorders; genitourinary disorders; neurodegenerative disorders

shipped in

wet ice

storage temp.

−20°C

General description

Creatine is present in vertebrates and helps to supply energy to muscle. In humans and animals, approximately half of creatine originates from food (mainly from fresh meat). Creatine supplementation has been investigated as a possible therapeutic approach for the treatment of muscular, neuromuscular, neurological and neurodegenerative diseases.

Features and Benefits

  • Linear detection range 4 to 1000 µM (colorimetric) or 0.5 to 50 µM (fluorometric).
  • Simple procedure allows for reliable quantitation of creatine within 30 minutes.

Suitability

The kit is suitable for creatine determination in biological samples (e.g. serum, plasma, urine, saliva etc).

Principle

The Creatine Assay Kit is based on enzymatic reactions leading to formation of a pink colored product. The optical density at 570 nm or fluorescence intensity at λEx = 530 nm/λEm = 590 nm is directly proportional to the creatine concentration in the sample.

related product

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service