Skip to Content
Merck
All Photos(1)

Key Documents

MAK463

Sigma-Aldrich

Hydroxyproline Assay Kit

sufficient for 100 colorimetric tests

Synonym(s):

Hydroxyproline Assay Kit (Perchlorate-free)

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$868.00

$868.00


Available to ship on14 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 KIT
$868.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

$868.00


Available to ship on14 April 2025Details


Request a Bulk Order

Quality Level

application(s)

cosmetics

detection method

colorimetric

relevant disease(s)

dermatological diseases; rheumatological diseases; neurological disorders; cancer; cardiovascular diseases

shipped in

ambient

storage temp.

2-8°C

General description

Hydroxyproline is a unique modified amino acid that is found exclusively in several animal proteins, the most prevalent of which is collagen. Collagen is a key structural protein of the extracellular matrix and is common in connective tissues due to its incredible strength. Because of its role as a key structural protein in animals, collagen is crucial for many medical applications, such as tissue transplantation and scaffolding for complex cell cultures. Collagen is also used in the beauty industry, as it can be supplemented to increase tissue rigidity, resulting in skin that appears more youthful.

Features and Benefits

  • Linear detection range 0.5 µg/mL to 50 µg/mL hydroxyproline
  • Assay uses an improved perchlorate-free chemistry

Suitability

The kit is suitable for hydroxyproline and collagen detection in biologic (e.g., serum, cells, urine, and tissue lysate) and cosmetic samples.

Principle

In the first step of this procedure, hydroxyproline in the sample is oxidized to a pyrrole ring. This compound then reacts with a dye reagent to yield a pink product that can be measured at 560 nm.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service