Skip to Content
Merck
All Photos(1)

Key Documents

MAK547

Sigma-Aldrich

High Sensitivity Beta-Hydroxybutyrate (Ketone Body) Assay Kit

sufficient for 200 fluorometric tests

Synonym(s):

High Sensitivity Beta-Hydroxybutyrate Assay Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$756.00

$756.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
$756.00

About This Item

UNSPSC Code:
41116104
NACRES:
NA.21

$756.00


Check Cart for Availability

Request a Bulk Order

application(s)

cosmetics
food and beverages

detection method

fluorometric

relevant disease(s)

endocrinological disorders, diabetes; gastrointestinal diseases

shipped in

dry ice

storage temp.

−20°C

General description

Ketone bodies are produced by the liver and used peripherally as an energy source when blood glucose levels drop. The two main ketone bodies are beta-hydroxybutyrate (β-HB) and acetoacetate, while acetone is the third abundant ketone body. Normally these two predominant ketone bodies are present in small amounts in the blood during fasting and prolonged exercise. In patients who have diabetes, alcohol or salicylate poisoning, hormone deficiency, childhood hypoglycemia and other acute disease states, large quantities of ketone bodies are found in the blood. The over-production and accumulation of ketone bodies in the blood (ketosis) can lead to pathological metabolic acidosis (ketoacidosis). Blood ketone testing methods that quantify β-HB, the predominant ketone body in the blood (approximately 75%) have been used for diagnosing and monitoring treatment of ketoacidosis.

​The Beta-Hydroxybutyrate Assay Kit offers a sensitive fluorescent assay for measuring Beta-HB levels in biological samples. This assay is based on an enzyme coupled reaction of β-HB, in which the product NADH can be specifically monitored by a fluorescent NADH sensor. The detection limit of this assay kit is 1.4 µM β-HB in a 100 µL reaction volume.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service