Skip to Content
Merck
All Photos(2)

Key Documents

MAK194

Sigma-Aldrich

Lipid (Oil Red O) Staining Kit

Sufficient for 2 96-well plates

Synonym(s):

Oil Red O Lipid Staining Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.25

relevant disease(s)

endocrinological disorders, diabetes; obesity; cardiovascular diseases

storage temp.

room temp

General description

Lipid droplets (LDs) are dynamic, ubiquitously present lipid-storage organelles, predominantly present in the adipocytes. Triglycerides, neutral lipids, and cholesterol esters stored in LDs are the largest sources of energy. The presence of excess LDs in adipocytes results in obesity and obesity-linked pathologies such as dyslipidemia and diabetes type.

Application

Lipid (Oil Red O) Staining Kit has been used to stain lipids[1] and adipocytes[2] in samples.

Suitability

This kit is suitable for staining neutral lipids in two 96 well plates, two 6 well plates, or four 100 mm culture dishes.

Principle

The Lipid (Oil Red O) Staining Kit is suitable for selective staining and detection of neutral lipids in cultured cells. Hematoxylin included in the kit stains the nuclei of the cells.

Kit Components Only

Product No.
Description

  • PBS

  • Formalin (10%)

  • Oil Red O

  • Hematoxylin

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CD36 mediates lipid accumulation in pancreatic beta cells under the duress of glucolipotoxic conditions: Novel roles of lysine deacetylases.
Khan S and Kowluru A
Biochemical and Biophysical Research Communications, 495(3), 2221-2226 (2018)
Kefeng Yang et al.
Xenotransplantation, 24(5) (2017-07-19)
Survival and longevity of xenotransplants depend on immune function and ability to integrate energy metabolism between cells from different species. However, mechanisms for interspecies cross talk in energy metabolism are not well understood. White adipose tissue stores energy and is
Yohei Nomura et al.
Frontiers in physiology, 12, 675724-675724 (2021-07-06)
We previously reported that histone deacetylase 6 (HDAC6) has an important role in endothelial cell (EC) function in vitro. However, whether HDAC6 plays a role in atherogenesis in vivo and the mechanism(s) that control HDAC6 activity/expression in response to atherogenic
Fatma Y Meligy et al.
Pharmaceutics, 14(7) (2022-07-28)
This study compared the cardioprotective action of mesenchymal stem cells (MSCs) and PUFAs in a rat model of gentamicin (GM)-induced cardiac degeneration. Male Wistar albino rats were randomized into four groups of eight rats each: group I (control group), group
Aldehyde dehydrogenase 1 a1 regulates energy metabolism in adipocytes from different species.
Yang K, et al.
Xenotransplantation, 24(5), e12318-e12318 (2017)

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. What are the methods for removing red stains from glassware from kit MAK194?

    1 answer
    1. For cleaning glassware from kit MAK194, it is suggested to rinse the glassware with ethanol and then scrub it with warm tap water and soap to remove the red staining caused by the Oil Red O solution. Following these steps with a standard wash in a lab dishwasher should yield optimal results. It's important to note that the longer the solution remains in the glassware, the more difficult it becomes to remove. Therefore, it is recommended to prepare special staining solutions in one-use plastic bottles to avoid this issue.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service