Skip to Content
Merck
All Photos(1)

Key Documents

MAK162

Sigma-Aldrich

Colorimetric Maleimide Assay Kit

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$461.00

$461.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
$461.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

$461.00


Check Cart for Availability

Request a Bulk Order

usage

sufficient for 100 colorimetric tests

detection method

colorimetric

relevant disease(s)

cancer

storage temp.

−20°C

General description

Maleimide is an unsaturated imide[1] used in protein conjugation reactions.[2] The direct measurement of maleimide moieties following conjugation reactions by spectrophotometric analysis at 302 nm is insensitive due to the small extinction coefficient (620M-1 cm-1) and complicated by protein absorbance at this wavelength.

Suitability

Suitable for quantifying maleimide groups

Principle

The colorimetric maleimide quantiation kit provides a simple and direct procedure for measuring maleimide groups. The sample is first reacted with a known amount of excess thiol and then the remaining unreacted thiol is assayed using 4,4′-DTDP (4,4′-Dithiodipyridine). The amount of maleimide is calculated as the difference between the initial amount of thiol and the amount of unreacted thiol after the complete reaction of all maleimide groups.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient site-specific labeling of proteins via cysteines.
Kim Y, et al.
Bioconjugate Chemistry, 19(3), 786-791 (2008)
Carbon Composites: Composites with Carbon Fibers, Nanofibers, and Nanotubes (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service