Skip to Content
Merck
All Photos(2)

Key Documents

MAK139

Sigma-Aldrich

Colorimetric Aldehyde Assay Kit

sufficient for 200 colorimetric tests

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$696.00

$696.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
$696.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

$696.00


Check Cart for Availability

Request a Bulk Order

usage

sufficient for 200 colorimetric tests

application(s)

environmental

detection method

colorimetric

relevant disease(s)

neurological disorders

storage temp.

−20°C

General description

Aldehydes are highly reactive molecules, which play a role in many physiological, pathological and commercial processes. They have been implicated as environmental pollutants, and in neurodegeneration and DNA damage. A method for rapid and accurate measurement of aldehydes is an important tool for biological and chemical research, the food industry, and environmental pollution surveillance.

Suitability

This kit is Suitable for the detection of aldehydes in a variety of samples, particularly when the samples are at a higher pH. When working with samples in a low pH, product number MAK140, Colorimetric Aldehyde Assay, is more suitable.

Principle

The Colorimetric Aldehyde Assay Kit provides a simple and direct procedure for measuring aldehydes in a variety of samples. Aldehydes reacts with a dye, resulting in a colorimetric (405 or 550 nm) product proportional to the amount of aldehyde present. The Colorimetric Aldehyde Assay Kit has a lower limit of detection of 1 nanomole of aldehyde in a 100 μL assay volume. This kit has been used for monitoring activities of oxidases that convert an amino group to an aldehyde group.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui Han et al.
Alcoholism, clinical and experimental research, 43(6), 1091-1102 (2019-03-26)
We previously developed enzyme nanoparticles (ENP) of alcohol metabolism. This study was to evaluate protective effects of facilitated removal of blood alcohol and/or acetaldehyde on anti-HIV drugs and alcohol-induced liver injuries. ENP were prepared for degrading alcohol completely (ENP1) or

Questions

  1. What is the composition of the aldehyde detection reagent for Product MAK139? Additionally, what is the concentration of this vial?

    1 answer
    1. The aldehyde sensor is composed of an aromatic hydrazine that rapidly reacts with an aldehyde to form a hydrozone. This hydrozone further reacts with a neighboring amino group to form a chromophore with maximum absorption at 550 nm. The concentration of aldehyde is proportional to the absorption intensity at 550 nm. The 2X working solution has a concentration of 32 mM.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service