Skip to Content
Merck
All Photos(1)

Documents

M6313

Sigma-Aldrich

Mimetic Green 1 Ligand Affinity Adsorbent

suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56

form

suspension

Quality Level

shipped in

wet ice

storage temp.

2-8°C

Application

Ligand affinity adsorbents are used in protein chromatography, affinity chromatography and dye resins. Ligand affinity adsorbents have been used to describe a simple protocol for the purification of Taq Pol from E. coli lysates as well as for the purification of other DNA polymerases.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B V Thage et al.
Journal of applied microbiology, 96(3), 593-602 (2004-02-14)
Purification and characterization of an aminotransferase (AT) specific for the degradation of branched-chain amino acids from Lactobacillus paracasei subsp. paracasei CHCC 2115. The purification protocol consisted of anion exchange chromatography, affinity chromatography and hydrophobic interaction chromatography. The enzyme was found
S C Woodcock et al.
The Biochemical journal, 313 ( Pt 2), 415-421 (1996-01-15)
CysG, also known as uroporphyrinogen III methylase and sirohaem synthase (CysG; EC 2.1.1.107), is a multifunctional enzyme that is able to transform uroporphyrinogen III into sirohaem via two S-adenosyl-L-methionine (AdoMet)-dependent transmethylations, an NAD(+)-dependent dehydrogenation and a ferrochelation. The apparent tight
Michael Kotik et al.
Biochimica et biophysica acta, 1760(2), 245-252 (2005-12-14)
Purification of a novel enantioselective epoxide hydrolase from Aspergillus niger M200 has been achieved using ammonium sulphate precipitation, ionic exchange, hydrophobic interaction, and size-exclusion chromatography, in conjunction with two additional chromatographic steps employing hydroxylapatite, and Mimetic Green. The enzyme was
J Ni et al.
The Journal of biological chemistry, 272(16), 10853-10858 (1997-04-18)
A new member of the human cystatin superfamily, called cystatin E, has been found by expressed sequence tag (EST) sequencing in amniotic cell and fetal skin epithelial cell cDNA libraries. The sequence of a full-length amniotic cell cDNA clone contained
Jeannie Horak et al.
Analytical and bioanalytical chemistry, 400(8), 2349-2359 (2010-12-22)
The new affinity-type Mimetic Ligand™ B14 was coupled with a 1,2-diaminoethane spacer (2LP) and a [1,2,3]-triazole spacer (TRZ) to three different support media. In addition to the agarose-based PuraBead and the polymethacrylate-type Fractogel, three new polymeric support media were introduced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service