Skip to Content
Merck
All Photos(1)

Key Documents

494542

Sigma-Aldrich

α-Cyclodextrin, sulfated sodium salt hydrate

Synonym(s):

α-Cyclodextrin hydrate sulfate sodium salt, alpha-cyclodextrin hydrate sulfate sodium

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$262.00

$262.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$262.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.22

$262.00


Check Cart for Availability

Request a Bulk Order

optical activity

[α]23/D +78°, c = 1 in H2O

mp

190 °C (dec.) (lit.)

SMILES string

[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[S](=O)(=O)([O-])O[C@@H]1[C@@H]2O[C@H]3[C@@H](O[C@@H](O[C@H]4[C@@H](O[C@@H](O[C@H]5[C@@H](O[C@@H](O[C@H]6[C@@H](O[C@@H](O[C@H]7[C@@H](O[C@@H](O[C@@H]([C@@H](O2)CO)[C@H]1O[S](=O)(=O)[O-])[C@H]([C@@H]7O)

InChI key

BXIKECRJPOGMAH-RQHIBQQSSA-E

General description

α-Cyclodextrin, sulfated sodium salt hydrate is a modified version of α-cyclodextrin, and is commonly used in the pharmaceutical industry to improve the solubility and bioavailability of poorly soluble drugs.

Application

α-Cyclodextrin, sulfated sodium salt hydrate can be used as a host molecule in the formation of inclusion complexes with the azo dye Methyl Yellow. It also enhance the stability of the hydrazone tautomer of Methyl Yellow in acidic medium.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service