Skip to Content
Merck
All Photos(2)

Key Documents

M4796

Sigma-Aldrich

(±)-α-Methyl-(4-carboxyphenyl)glycine

Synonym(s):

(±)-MCPG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11NO4
CAS Number:
Molecular Weight:
209.20
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

SMILES string

CC(N)(C(O)=O)c1ccc(cc1)C(O)=O

InChI

1S/C10H11NO4/c1-10(11,9(14)15)7-4-2-6(3-5-7)8(12)13/h2-5H,11H2,1H3,(H,12,13)(H,14,15)

InChI key

DNCAZYRLRMTVSF-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Competitive antagonist at Group 1 (mGluR1 and mGluR5) and Group 2 (mGluR2 and mGluR3) metabotropic glutamate receptors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hilary Gerstein et al.
Neurobiology of learning and memory, 97(1), 17-29 (2011-09-29)
Homer1 belongs to a family of scaffolding proteins that interact with various post-synaptic density proteins including group I metabotropic glutamate receptors (mGluR1/5). Previous research in our laboratory implicates the Homer1c isoform in spatial learning. Homer1 knockout mice (H1-KO) display cognitive
D D Schoepp et al.
Neuropharmacology, 36(1), 1-11 (1997-01-01)
The novel compound LY354740 is a conformationally constrained analog of glutamate, which was designed for interaction at metabotropic glutamate (mGlu) receptors. In this paper the selectivity of LY354740 for recombinant human mGlu receptor subtypes expressed in non-neuronal (RGT) cells is
Ana C Takakura et al.
Experimental physiology, 96(10), 989-999 (2011-07-12)
In the present study, we evaluated the role of glutamatergic mechanisms in the retrotrapezoid nucleus (RTN) in changes of splanchnic sympathetic nerve discharge (sSND) and phrenic nerve discharge (PND) elicited by central and peripheral chemoreceptor activation. Mean arterial pressure (MAP)
L S Thomas et al.
Neuropharmacology, 39(9), 1554-1566 (2000-06-16)
In the present study we have examined the role of presynaptic group I metabotropic glutamate (mGlu) receptors in the control of neuronal glutamate release using rat forebrain slices pre-loaded with [(3)H]D-aspartate. We have also addressed the question of which group
A M Rao et al.
Neuroscience letters, 293(1), 1-4 (2000-11-07)
Stimulation of group I metabotropic glutamate receptors (mGluR 1 and 5) activates G-protein coupled-phospholipase C (PLC) to release 1,2-diacylglycerol (DAG) and arachidonic acid (ArAc). To elucidate the role of group I mGluR, we tested the effects of (S)-alpha-methyl-4-carboxy-phenylglycine (MCPG, mGluR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service