Key Documents
L8663
Sigma-Aldrich
Director-Ready pc-myc-CMV-2
About This Item
UNSPSC Code:
12171500
NACRES:
NA.85
Pricing and availability is not currently available.
storage temp.
−20°C
General description
The Director-Ready� pc-myc-CMV2 Expression Vector is de-phosphorylated after Hind III/Bgl II digestion and is ready for use with the Director Universal Cloning System. pc-myc-CMV2 is for transient, cytoplasmic expression of N-terminal c-myc fusion proteins in mammalian cells.
Legal Information
Director-Ready is a trademark of Sigma-Aldrich Co. LLC
pc-myc-CMV-2 is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)