Skip to Content
Merck
All Photos(1)

Key Documents

M042080

Sigma-Aldrich

Biotin Phosphoramidite

configured for PerkinElmer, configured for Polygen

Synonym(s):

Biotin amidite

Sign Into View Organizational & Contract Pricing

Select a Size

0.1 G
$217.00
0.25 G
$434.00

$217.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
0.1 G
$217.00
0.25 G
$434.00

About This Item

Empirical Formula (Hill Notation):
C46H64N5O8PS
CAS Number:
Molecular Weight:
878.07
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21

$217.00


Check Cart for Availability

Request a Bulk Order

product line

Proligo Reagents

Assay

≥96% (31P-NMR)

suitability

conforms to structure for H-NMR

compatibility

configured for PerkinElmer
configured for Polygen

storage temp.

−20°C

SMILES string

COc1ccc(cc1)C(N2C3CS[C@@H](CCCCC(=O)NCCOCCOCCOP(OCCC#N)N(C(C)C)C(C)C)C3NC2=O)(c4ccccc4)c5ccc(OC)cc5

InChI

1S/C46H64N5O8PS/c1-34(2)51(35(3)4)60(58-27-12-25-47)59-32-31-57-30-29-56-28-26-48-43(52)16-11-10-15-42-44-41(33-61-42)50(45(53)49-44)46(36-13-8-7-9-14-36,37-17-21-39(54-5)22-18-37)38-19-23-40(55-6)24-20-38/h7-9,13-14,17-24,34-35,41-42,44H,10-12,15-16,26-33H2,1-6H3,(H,48,52)(H,49,53)/t41-,42-,44-,60?/m0/s1

InChI key

WSDQYLGSIUJJRU-ZMKGMYJVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Biotin phosphoramidite is a reagent used to directly incorporate a biotin label at the 5′ hydroxy group of the terminal nucleoside base through a p-aminophenethyl spacer group during oligonucleotide synthesis.

The photocleavable biotin phosphoramidite reagent is compatible with automated DNA/RNA synthesizers using phosphoramidite chemistry. The length of the p-aminophenethyl spacer group allows streptavidin affinity purification of the oligonucleotide by reverse-phase HPLC. Biotinlabeling is compatible with PCR and most hybridization techniques.Key Features of Biotin Phosphoramidite
  • Behaves like any standard phosphoramidite on the DNA synthesizer
  • Readily soluble in acetonitrile
  • Biotin compound has a dimethoxytrityl (DMT) group on the ring structure,which allows coupling-yield determination and trityl-selective purification• A long, water-compatible linker arm ensures maximum sensitivity
  • Provides a coupling efficiency of at least 95%The product is configured for Expedite and PolyGen® Synthesizers.

Application

Biotin Phosphoramidite is suitable for the synthesis of 5′-biotinylated oligonucleotides whose applications are seen in ligase-mediated gene detection, direct dideoxy sequencing following PCR, and non-radioactive sequencing of DNA.

Legal Information

Expedite is a trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries
PolyGen is a registered trademark of PolyGen GmbH.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service