Skip to Content
Merck
All Photos(3)

Key Documents

L3520

Sigma-Aldrich

D-Lactitol monohydrate

≥99% (HPLC)

Synonym(s):

4-O-β-D-Galactopyranosyl-D-glucitol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$128.00
5 G
$454.00

$128.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$128.00
5 G
$454.00

About This Item

Empirical Formula (Hill Notation):
C12H24O11 · H2O
CAS Number:
Molecular Weight:
362.33
Beilstein:
4218679
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

$128.00


Check Cart for Availability

Request a Bulk Order

Assay

≥99% (HPLC)

form

powder

sweetness

0.4 × sucrose

color

white

mp

95-98 °C (lit.)

solubility

water: 50 mg/mL, clear, colorless

SMILES string

[H]O[H].OC[C@H](O)[C@@H](O)[C@H](O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O)[C@H](O)CO

InChI

1S/C12H24O11.H2O/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12;/h4-21H,1-3H2;1H2/t4-,5+,6+,7+,8-,9-,10+,11+,12-;/m0./s1

InChI key

LXMBXZRLTPSWCR-XBLONOLSSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Sugar alcohol sweet tastant detectable by humans.

Other Notes

To gain a comprehensive understanding of our extensive range of Sugar alcohols for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service