Skip to Content
Merck
All Photos(1)

Key Documents

PHR1769

Supelco

Isomalt

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Isomalt, 6-O-α-D-Glucopyranosyl-D-glucitol mixed with 1-O-α-D-glucopyranosyl-D-mannitol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$176.00

$176.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
1 G
$176.00

About This Item

Empirical Formula (Hill Notation):
C12H24O11
CAS Number:
Molecular Weight:
344.31
UNSPSC Code:
41116107
NACRES:
NA.24

$176.00


In StockDetails


Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. I0465000
traceable to USP 1349626

API family

isomalt

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O1[C@@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)OC[C@@H](O)[C@@H](O)[C@H](O)C(O)CO

InChI

1S/C12H24O11/c13-1-4(15)7(17)8(18)5(16)3-22-12-11(21)10(20)9(19)6(2-14)23-12/h4-21H,1-3H2/t4?,5-,6-,7-,8-,9-,10+,11-,12+/m1/s1

InChI key

SERLAGPUMNYUCK-BLEZHGCXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isomalt is a disaccharide alcohol that is commonly used as an excipient in pharmaceutical formulations because of its beneficial characteristics such as the taste, mouth feel, low calorie content, acariogenicity, suitability to diabetics, high stability and low hygroscopicity.[1]
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Isomalt may be co-extruded with paracetamol or hydrochlorothiazide in order to improve the tabletting properties and subsequently, the determination of the analyte in pharmaceutical formulations may be carried out using high-performance liquid chromatography (HPLC).[1] It may also be used as an excipient for meloxicam active pharmaceutical ingredient (API) and the determination of the analyte in pharmaceutical formulations can be carried out using near infra-red (NIR) chemometric method.[2]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC0143 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantification of meloxicam and excipients on intact tablets by near infrared spectrometry and chemometry
TOMUT? I, et al.
Farmacia, 58(5), 559-571 (2010)
F Ndindayino et al.
International journal of pharmaceutics, 235(1-2), 159-168 (2002-03-07)
Isomalt, a disaccharide alcohol was co-extruded with paracetamol or hydrochlorothiazide (HCT) in order to improve its tabletting properties. After extrusion, isomalt was transformed into an amorphous form, while paracetamol remained crystalline. Hot stage microscopy showed that HCT was amorphous in

Questions

  1. I would like to buy isomalt standard for HPLC?  What is the purity of PHR1769 Isomalt?  Is it possible to use this?  Thank you. 

    1 answer
    1. Yes, this product is tested and suitable for use in HPLC. The purity of this product is approximately 95% as reported in the lot-specific Certificate of Analysis. Please see the link below to review a sample or lot specific Certificate:
      https://www.sigmaaldrich.com/product/sial/phr1769#product-documentation

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service