Skip to Content
Merck
All Photos(1)

Key Documents

I0635

Sigma-Aldrich

4,4′-Isopropylidenediphenol

Synonym(s):

2,2-Bis(4-hydroxyphenyl)propane, Bisphenol A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16O2
CAS Number:
Molecular Weight:
228.29
Beilstein:
1107700
EC Number:
MDL number:
UNSPSC Code:
12000000

SMILES string

CC(C)(c1ccc(O)cc1)c2ccc(O)cc2

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

440.6 °F - closed cup

Flash Point(C)

227 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Séverine H Collet et al.
Toxicology and applied pharmacology, 284(3), 323-329 (2015-03-12)
The investigation of interspecies differences in bisphenol A (BPA) pharmacokinetics (PK) may be useful for translating findings from animal studies to humans, identifying major processes involved in BPA clearance mechanisms, and predicting BPA PK parameters in man. For the first
Chandrew Rajakumar et al.
Reproductive toxicology (Elmsford, N.Y.), 53, 39-44 (2015-03-19)
This study examined the effect of bisphenol A (BPA) on human placental gene expression using primary trophoblast cells as an in vitro model system. Trophoblast cells were isolated from human placentas at term, cultured and then exposed to environmentally relevant
Christopher Elam et al.
European journal of medicinal chemistry, 46(5), 1512-1523 (2011-03-01)
Two screening protocols based on recursive partitioning and computational ligand docking methodologies, respectively, were employed for virtual screens of a compound library with 345,000 entries for novel inhibitors of the enzyme sarco/endoplasmic reticulum calcium ATPase (SERCA), a potential target for
Marie Tremblay-Franco et al.
PloS one, 10(10), e0141698-e0141698 (2015-10-31)
Along with the well-established effects on fertility and fecundity, perinatal exposure to endocrine disrupting chemicals, and notably to xeno-estrogens, is strongly suspected of modulating general metabolism. The metabolism of a perinatally exposed individual may be durably altered leading to a
Bhavini B Patel et al.
Toxicological sciences : an official journal of the Society of Toxicology, 146(1), 101-115 (2015-04-12)
Estrogenic compounds such as bisphenol A (BPA) leach from plastics into food and beverage containers. Increased BPA exposure has been correlated with increased cardiovascular disease. To test the hypothesis that increased BPA exposure reduces cardiovascular remodeling, we chronically exposed C57bl/6n

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service