Merck
  • Chronic Exposure to Bisphenol A Reduces Successful Cardiac Remodeling After an Experimental Myocardial Infarction in Male C57bl/6n Mice.
Chronic Exposure to Bisphenol A Reduces Successful Cardiac Remodeling After an Experimental Myocardial Infarction in Male C57bl/6n Mice.
Toxicological sciences : an official journal of the Society of Toxicology (2015-04-12)
Bhavini B Patel, Amanda Kasneci, Alicia M Bolt, Vanessa Di Lalla, Massimo R Di Iorio, Mohamad Raad, Koren K Mann, Lorraine E Chalifour
ABSTRACT
Estrogenic compounds such as bisphenol A (BPA) leach from plastics into food and beverage containers. Increased BPA exposure has been correlated with increased cardiovascular disease. To test the hypothesis that increased BPA exposure reduces cardiovascular remodeling, we chronically exposed C57bl/6n male mice to BPA and performed a myocardial infarction (MI). We measured cardiac function, as well as myeloid and cardiac fibroblast accumulation and activity. We found increased early death as well as increased cardiac dilation and reduced cardiac function in surviving BPA-exposed mice. Matrix metalloproteinase-2 (MMP2) protein and activity were increased 1.5-fold in BPA-exposed heart. BPA-exposed mice had similar neutrophil infiltration; however, monocyte and macrophage (MΦ) infiltration into the ischemic area was 5-fold greater than VEH mice potentially due to a 2-fold increase in monocyte chemoattractant protein-1. Monocyte and MΦ exposure to BPA in vitro in primary bone marrow cultures or in isolated peritoneal MΦ increased polarization to an activated MΦ, increased MMP2 and MMP9 expression 2-fold and activity 3-fold, and increased uptake of microspheres 3-fold. Cardiac fibroblasts (CF) differentiate to α-smooth muscle actin (αSMA) expressing myofibroblasts, migrate to the ischemic area and secrete collagen to strengthen the scar. Collagen and αSMA expression were reduced 50% in BPA-exposed hearts. Chronic in vivo or continuous in vitro BPA exposure ablated transforming growth factor beta-mediated differentiation of CF, reduced αSMA expression 50% and reduced migration 40% yet increased secreted MMP2 activity 2-fold. We conclude that chronic BPA exposure reduces the ability to successfully remodel after an MI by increasing MΦ-based inflammation and reducing myofibroblast repair function.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O