Skip to Content
Merck
All Photos(1)

Documents

HUMANPBMC-0002200

Sigma-Aldrich

Frozen Human Rheumatoid Arthritis PBMC

human peripheral blood (mononuclear cells)

Synonym(s):

Immune Cells, Peripheral Blood Mononuclear Cells, RA PBMC, Rheumatoid Arthritis PBMC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

Frozen Human Rheumatoid Arthritis PBMC, 20 Million Cells/per vial

biological source

human peripheral blood (mononuclear cells)

Quality Level

form

frozen liquid

packaging

vial of 20 Million cells

manufacturer/tradename

BioIVT

technique(s)

cell culture | mammalian: suitable

color

cloudy

storage temp.

-140 to -196°C

General description

Lot specific orders are not able to be placed throughthe web. Contact your local sales rep for more details.


We are pleased to offer these products from BioIVT.Offering isolated Peripheral Blood Mononuclear Cells(PBMCs) from whole blood procured from IRB-consented donors with clinicallydefined Rheumatoid Arthritis.

Peripheral Blood MononuclearCells (PBMCs) are a subset of leukocytes (or White Blood Cell, WBC), which areintegral components of the body′simmune system that fight disease.

PBMCs consist of a mixtureof lymphocytes (T cells, B cells, and NK cells) and monocytes and are isolatedfrom peripheral whole blood via density gradient centrifugation and red bloodcell lysis.

Standard and custom clinical data collection andconfirmation ensures proper corresponding disease severity and treatmenthistory (prescription and over the counter).

Clinical datamay include: Current medical treatment, CDAI Score, CRP Score, HLA, RF, CCPOurreadily available

PBMCs are provided with clinical data thathas been collected following a strict and standardized clinical data process.All specimens are reviewed for IRB approval.

Packaging

20 Million Cells/per vial

Other Notes

Thaw before use.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service