Skip to Content
Merck
All Photos(1)

Key Documents

HUMANPBMC-0002180

Sigma-Aldrich

Frozen Human Systemic Lupus Erythematosus (SLE) PBMC

human peripheral blood (mononuclear cells)

Synonym(s):

Immune Cells, Peripheral Blood Mononuclear Cells, Systemic Lupus Erythematosus PBMC, UC PBMC

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
$1,210.00

$1,210.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 VIAL
$1,210.00

About This Item

UNSPSC Code:
41106514

$1,210.00


Check Cart for Availability

Request a Bulk Order

Product Name

Frozen Human Systemic Lupus Erythematosus (SLE) PBMC, 20 Million Cells/per vial

biological source

human peripheral blood (mononuclear cells)

Quality Level

form

frozen liquid

packaging

vial of 20 Million cells

manufacturer/tradename

BioIVT

technique(s)

cell culture | mammalian: suitable

color

cloudy

storage temp.

-140 to -196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

We are pleased to offer these products from BioIVT.

Offering isolated Peripheral Blood Mononuclear Cells (PBMCs) from whole blood procured from IRB-consented donors with clinically defined Systemic Lupus Erythematosus (SLE).

Peripheral Blood Mononuclear Cells (PBMCs) are a subset of leukocytes (or White Blood Cell, WBC), which are integral components of the body′s immune system that fight disease.

PBMCs consist of a mixture of lymphocytes (T cells, B cells, and NK cells) and monocytes and are isolated from peripheral whole blood via density gradient centrifugation and red blood cell lysis.

Standard and custom clinical data collection and confirmation ensures proper corresponding disease severity and treatment history.

Clinical data may include: Medication (current and historical), Disease severity, Standard blood chemistry results, dsDNA results, C3/C4 results, ANA results

PBMCs are provided with clinical data that has been collected following a strict and standardized clinical data process. All specimens are reviewed for IRB approval.

Packaging

20 Million Cells/per vial

Other Notes

Thaw before use.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service