HUMANCD14-0002080
Frozen Human Normal CD14+ Monocytes
human peripheral blood (monocytes)
Synonym(s):
CD14+ Cells, Immune Cells, Monocytes
About This Item
Recommended Products
product name
Frozen Human Normal CD14+ Monocytes, 20 Million Cells/per vial
biological source
human peripheral blood (monocytes)
Quality Level
form
frozen liquid
packaging
vial of 20 Million cells
manufacturer/tradename
BioIVT
technique(s)
cell culture | mammalian: suitable
color
cloudy
storage temp.
-140 to -196°C
General description
Monocytes are a subset of leukocytes, or white blood cells, found in peripheral blood and can migrate into tissue to differentiate into macrophages or myeloid lineage dendritic cells. CD14+ Monocytes are isolated from peripheral blood and separated from mononuclear cells via immunomagnetic separation.
Used for a variety of applications, monocytes are frequently utilized in drug discovery/development, assay validation/development, monocyte activation tests, and other immunology applications.
Packaging
Other Notes
Disclaimer
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
FAQs about BioIVT cell lines and culture, including cryopreserved PBMCs, BMMCs, and immune cell lines.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service