Skip to Content
Merck
All Photos(2)

Key Documents

HT110316

Sigma-Aldrich

Eosin Y Solution, Alcoholic, with Phloxine

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
$115.00

$115.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 ML
$115.00

About This Item

MDL number:
UNSPSC Code:
41116124
NACRES:
NA.47

$115.00


Check Cart for Availability

Request a Bulk Order

form

solution

Quality Level

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

application(s)

hematology
histology

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

General purpose cytoplasmic counterstain. Used with hematoxylin and eosin staining.

Other Notes

Certified Eosin Y, 0.1% (w/v) and certified phloxine B, 0.1% (w/v) in acidified ethanol

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Sens. 1 - STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bertrand Bryche et al.
Brain, behavior, and immunity, 79, 274-283 (2019-02-19)
At the interface of the environment and the nervous system, the olfactory mucosa (OM) is a privileged pathway for environmental toxicants and pathogens towards the central nervous system. The OM is known to produce antimicrobial and immunological components but the
Agnes Bonnet et al.
BMC genomics, 12, 417-417 (2011-08-20)
Successful achievement of early folliculogenesis is crucial for female reproductive function. The process is finely regulated by cell-cell interactions and by the coordinated expression of genes in both the oocyte and in granulosa cells. Despite many studies, little is known
Eva Bártová et al.
Histochemistry and cell biology, 145(4), 447-461 (2016-01-23)
We studied the histone signature of embryonic and adult brains to strengthen existing evidence of the importance of the histone code in mouse brain development. We analyzed the levels and distribution patterns of H3K9me1, H3K9me2, H3K9me3, and HP1β in both
Yi Chen et al.
Scientific reports, 9(1), 5624-5624 (2019-04-06)
The tumor microenvironment can be spatially heterogenous, which makes it challenging to fully characterize with standard 2D histology-based methods. In this study, we determined the feasibility of a CLARITY tissue-processing approach to analyze biopsies from breast cancer patients. Formalin-fixed human
Tae-Muk Kim et al.
Journal of ethnopharmacology, 253, 112082-112082 (2019-07-17)
Eriobotrya japonica leaves has a very long history of medicinal use as an anti-inflammatory and antitussive agent for bronchial inflammation. The aim of this study was to evaluate the anti-inflammatory activities of Eriobotrya japonica (EJ) leaf water extract in an

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service