Skip to Content
Merck
All Photos(1)

Key Documents

1.17081

Sigma-Aldrich

Eosin Y solution 1%, alcoholic

for microscopy

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$219.00

$219.00


Available to ship on03 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 L
$219.00

About This Item

UNSPSC Code:
41116124
NACRES:
NA.41

$219.00


Available to ship on03 April 2025Details


Request a Bulk Order

Quality Level

form

liquid

IVD

for in vitro diagnostic use

technique(s)

microbe id | staining: suitable

transition temp

flash point 19 °C

density

0.892 g/cm3 at 20 °C

application(s)

clinical testing
diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

Eosin Y solution 1%, alcoholic - for microscopy is used for human-medical cell diagnosis and serves the histological and clinico-cytological investigation of sample material of human origin in histological and clinico-cytological specimen materials, for example histological sections of e. g. the kidney, muscle tissue, heart, or lung. The hematoxylin and eosin (H&E) staining method is the method most frequently used for the staining of histology material. The staining mechanism behind the H&E staining is a physico-chemical process. In the first step, the positively charged nuclear dye (hematoxylin) binds to the negatively charged phosphate groups of the nucleic acid of the cell nucleus. The nuclei will be dyed dark blue to dark violet. The second step is the counterstaining with negatively charged anionic Eosin Y, a xanthene dye. Eosin binds to the positively charged plasma proteins. Cytoplasma and intercellular substances are stained pink to red, while erythrocytes will appear with yellow to orange color.
One 500ml bottle of the stain provides 1250 stainings. For more details, please see instructions for use (IFU). This product is registered as IVD and CE marked.

Analysis Note

Suitability for microscopy (Tissue section): passes test
Nuclei: dark blue to dark violet
Cytoplasm: pink to red
intercellular substances: pink to red
Erythrocytes: yellow to orange

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

66.2 °F

Flash Point(C)

19 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mare Mechelinck et al.
PloS one, 14(12), e0226430-e0226430 (2019-12-14)
A simple but reliable and safe anaesthetic procedure is required for surgical interventions in small rodents. Combined ketamine and xylazine injections are often used in rats for less invasive surgery, possibly with spontaneous breathing and without airway management. However, there

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service