Skip to Content
Merck
All Photos(2)

Documents

HT1014

Sigma-Aldrich

Hydroquinone tablets

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116124
PubChem Substance ID:
NACRES:
NA.47

form

powder

Quality Level

shelf life

expiry date on the label

pH

3.7

application(s)

hematology
histology

storage temp.

2-8°C

Application

Intended for use in the Silver Stain (Steiner-Steiner) Kit.

Components

Hydroquinone, 0.5 g each, with excipients.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

329.0 °F - closed cup

Flash Point(C)

165 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service