Skip to Content
Merck
All Photos(2)

Key Documents

HS8610A

Laboratory Notebook

100 pages, grid, black

Synonym(s):

experiment notebook, lab notebook

Sign Into View Organizational & Contract Pricing

Select a Size

12 EA
$726.00

$726.00


Check Cart for Availability


Select a Size

Change View
12 EA
$726.00

About This Item

UNSPSC Code:
14111514
NACRES:
NB.35

$726.00


Check Cart for Availability

feature

grid

packaging

pk of 12

manufacturer/tradename

Heathrow Scientific HS8610A

L × W

218 × 281 mm

color

black

Looking for similar products? Visit Product Comparison Guide

General description

Use these notebooks to keep information safe and well organized. Record research data, experimental procedures, and design ideas.

Features and Benefits

  • numbered, acid-free, pages include spaces for title, project number, book number, date, and witness signature
  • lined notebooks feature college-ruled pages for easy report writing
  • covers for all versions are waterproof and chemical resistant
  • all notebooks contain periodic and conversion tables in the back


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service