Key Documents
HS120250
Wide mouth safety labelled wash bottle
capacity 500 mL, label, Acetone, self venting, PE-LD, acetone
Select a Size
1 EACH
$84.30
$84.30
Available to ship on17 April 2025Details
Select a Size
Change View
1 EACH
$84.30
About This Item
UNSPSC Code:
41103209
NACRES:
NB.43
$84.30
Available to ship on17 April 2025Details
material
PE-LD
low-density polyethylene bottle
polypropylene cap
red bottle
wide-mouth bottle
feature
label Acetone
vented
with cap
packaging
pack of 6 ea
manufacturer/tradename
Heathrow Scientific HS120250
capacity
500 mL
General description
Clear and informative product safety information.
The information provided on the front of each bottle is specific to the product contained and includes the following — the Chemical Abstract Service (CAS) No., National Fire Protection Association (NFPA) codes and symbols, Protective Equipment Symbol (PES), Target Organ Information and Hazard Codes. You also have the option of using other liquids within the bottles and creating the appropriate safety information on the outside of the bottle by using the ‘Self Labeling′ versions. The closure and stem are molded in one piece to make them leak-proof. The angled neck and tip provide a uniform stream of liquid and the tip can be cut back if required to increase the flow.
  • Wide bottle mouth makes filling easier
  • Non-metallic vent that prevents the risk of solvent leakage
  • With the exception of the bottle designed for Sodium Hypochlorite, which is a white LDPE bottle for light protection and has a HDPE closure, the bottles are made from LDPE, with a PP closure
  • Six named chemicals, a self labeling and assorted pack of bottles to choose from
  • 500 mL bottle size
  • Supplied in packs of 6
Self Labeling Bottles must only be used with chemicals that are safe and compatible with LDPE
Choose from one of the most recent versions:
Certificates of Analysis (COA)