Key Documents
HS120201
Microtube Storage Box
blue polypropylene, pk of 5, Holds 100 x 1.5-2.0 mL tubes
Synonym(s):
tube rack, tube storage
Select a Size
5 EA
$127.00
$127.00
Available to ship on24 April 2025Details
Select a Size
Change View
5 EA
$127.00
About This Item
UNSPSC Code:
41123300
NACRES:
NB.22
$127.00
Available to ship on24 April 2025Details
material
blue polypropylene
feature
Holds 100 x 1.5-2.0 mL tubes
packaging
pk of 5
manufacturer/tradename
Heathrow Scientific HS120201
well diam.
12.3 mm
Features and Benefits
  • for larger storage requirements of 1.5-2.0 mL microtubes or cryogenic vials
  • easy-open hinged lid with snap clasp
  • autoclavable
  • designed for quick visual identification
  • uniform wells with location indentification
  • tube capacity is indicated on the outside of the box
  • translucent vibrant colors with frosted labeling area
  • debossed with a distinctive wave design
Choose from one of the most recent versions:
Certificates of Analysis (COA)