Key Documents
Z207187
Sigma-Aldrich
Broken-glass disposal boxes
Benchtop
Synonym(s):
benchtop waste glass box, laboratory waste glass box
Select a Size
6 EA
$182.00
$182.00
Available to ship on23 April 2025Details
Select a Size
Change View
6 EA
$182.00
About This Item
UNSPSC Code:
41123300
NACRES:
NB.35
$182.00
Available to ship on23 April 2025Details
material
cardboard
feature
Holds broken glass
packaging
pkg of 6 ea
manufacturer/tradename
Bel-Art F24653-0002
availability
not available in EU
W × L × H
8 in. × 8 in. × 10 in.
General description
Corrugated cardboard receptacle supplied with 2 mil PE bag. When full, safety cap is pulled into place and entire unit is ready for disposal. Printed in four languages (English, French, Spanish, and German). Shipped flat.
Choose from one of the most recent versions:
Certificates of Analysis (COA)