HS10323
Biohazard disposal bags
H × W × thickness 838 mm × 635 mm × 0.04 mm, red polypropylene (with biohazard warning symbol & precautions in English, Spanish, & French)
Synonym(s):
autoclave waste disposal, biohaz waste bag, biohazardous disposal bags, waste bags
Select a Size
$485.00
Available to ship on07 April 2025Details
Select a Size
About This Item
$485.00
Available to ship on07 April 2025Details
Recommended Products
material
red polypropylene (with biohazard warning symbol & precautions in English, Spanish, & French)
feature
label
packaging
pkg of 200 ea
manufacturer/tradename
Heathrow Scientific HS10323
H × W × thickness
838 mm × 635 mm × 0.04 mm
Related Categories
General description
A range of biohazard collection bags printed with biohazard warning symbol and precautions in English, Spanish, and French
- Includes an indicator patch that darkens when autoclaved
- Autoclavable to 250°F (121°C)
- Disposable
- Epoxy coated steel wire holder available HS10044H
Features and Benefits
- Two thicknesses of polypropylene are available
- An indicator patch that darkens when autoclaved is included
- Autoclavable to 250°F (121°C)
- Disposable
- An epoxy-coated steel wire holder is available (HS10044H).
Choose from one of the most recent versions:
Certificates of Analysis (COA)