Skip to Content
Merck
All Photos(1)

Key Documents

HS10321

Biohazard disposal bags

H × W × thickness 483 mm × 356 mm × 0.04 mm, red polypropylene (with biohazard warning symbol & precautions in English, Spanish, & French)

Synonym(s):

autoclave waste disposal, biohaz waste bag, biohazardous disposal bags, waste bags

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$132.00

$132.00


Check Cart for Availability


Select a Size

Change View
1 EACH
$132.00

About This Item

UNSPSC Code:
41106514
NACRES:
NB.17

$132.00


Check Cart for Availability

material

red polypropylene (with biohazard warning symbol & precautions in English, Spanish, & French)

feature

label

packaging

pkg of 200 ea

manufacturer/tradename

Heathrow Scientific HS10321

H × W × thickness

483 mm × 356 mm × 0.04 mm

Looking for similar products? Visit Product Comparison Guide

General description

Biohazard disposal bags are printed with biohazard warning symbols. A wide range of biohazard collection bags with sizes to suit all needs are available for personal or general lab use.
For personal or general lab use, sizes to suit all needs
A range of biohazard collection bags printed with biohazard warning symbol and precautions in English, Spanish, and French
  • Includes an indicator patch that darkens when autoclaved
  • Autoclavable to 250°F (121°C)
  • Disposable
  • Epoxy coated steel wire holder available HS10044H

Features and Benefits

  • Two thicknesses of polypropylene are available
  • An indicator patch that darkens when autoclaved is included
  • Autoclavable to 250°F (121°C)
  • Disposable
  • An epoxy-coated steel wire holder is available (HS10044H).

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. how many per box ?

    1 answer
    1. As mentioned in the 'PROPERTIES' section, this product is supplied as a package of 200 biohazard disposal bags.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service