Skip to Content
Merck
All Photos(3)

Key Documents

HPA022021

Sigma-Aldrich

Anti-EFTUD2 antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-SNRNP116, Anti-Snrp116, Anti-Snu114, Anti-U5-116KD

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Estimated to ship on12 May 2025



Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Estimated to ship on12 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL

immunogen sequence

DDGVQFHAFGRVLSGTIHAGQPVKVLGENYTLEDEEDSQICTVGRLWISVARYHIEVNRVPAGNWVLIEGVDQPIVKTATITEPRGNEEAQIFRPLKFNTTSVIKIAVEPV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EFTUD2(9343)

General description

EFTUD2 (Elongation factor Tu GTP binding domain containing 2) is a member of the U5 small nuclear ribonucleoprotein particle (snRNP) located on chromosome region 17q21.31. It encodes a small GTPase (116kDa) component of the major spliceosome. It consists of a GTP-binding domain and several other conserved domains homologous to the translational elongation factor EF-2. Its N-terminal domain has an acidic domain.

Immunogen

116 kDa U5 small nuclear ribonucleoprotein component recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

EFTUD2 (Elongation factor Tu GTP binding domain containing 2) acts as a component of the spliceosome complex during translocation of mRNA on the ribosome. It produces mature mRNAs by binding to GTP during the processing of precursor mRNAs. Haploinsufficient genetic mutation in EFTUD2 causes a sporadic malformation syndrome, mandibulofacial dysostosis with microcephaly (MFDM), characterized with severe craniofacial abnormalities, microcephaly, growth delay, hearing loss, cleft palate, choanal atresia and dysmorphic features.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75576

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Lei et al.
Nucleic acids research, 45(6), 3422-3436 (2016-12-03)
Haploinsufficiency of EFTUD2 (Elongation Factor Tu GTP Binding Domain Containing 2) is linked to human mandibulofacial dysostosis, Guion-Almeida type (MFDGA), but the underlying cellular and molecular mechanisms remain to be addressed. We report here the isolation, cloning and functional analysis
Margaret P Adam et al.
GeneReviews(?), 2014 Jul 03 (2014-07-08)
Mandibulofacial dysostosis with microcephaly (MFDM) is characterized by malar and mandibular hypoplasia; microcephaly (congenital or postnatal onset); malformations of the pinna, auditory canal, and/or middle ear (ossicles and semi-circular canals) with associated conductive hearing loss; and distinctive facial features (metopic
Daphné Lehalle et al.
Human mutation, 35(4), 478-485 (2014-01-29)
Mandibulofacial dysostosis, Guion-Almeida type (MFDGA) is a recently delineated multiple congenital anomalies/mental retardation syndrome characterized by the association of mandibulofacial dysostosis (MFD) with external ear malformations, hearing loss, cleft palate, choanal atresia, microcephaly, intellectual disability, oesophageal atresia (OA), congenital heart
T Achsel et al.
Molecular and cellular biology, 18(11), 6756-6766 (1998-10-17)
The human small nuclear ribonucleoprotein (snRNP) U5 is biochemically the most complex of the snRNP particles, containing not only the Sm core proteins but also 10 particle-specific proteins. Several of these proteins have sequence motifs which suggest that they participate
S K Gandomi et al.
Clinical genetics, 87(1), 80-84 (2013-11-26)
Mandibulofacial dysostosis with microcephaly (MFDM) is a sporadic malformation syndrome with severe craniofacial abnormalities, microcephaly, developmental delay, and dysmorphic features. Most cases of clinically diagnosed MFDM remain genetically unexplained, and to the best of our knowledge a total of 35

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service