Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200725

Sigma-Aldrich

Anti-Splicing Factor SC-35 antibody, Mouse monoclonal

clone SC-35, purified from hybridoma cell culture

Synonym(s):

Anti-PR264, Anti-SFRS2, Anti-SFRS2A, Anti-SRp30b, Anti-Splicing component 35 kDa (SC35), Anti-serine/arginine-rich splicing factor 2 (SRSF2)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$830.00

$830.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$830.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$830.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

SC-35, monoclonal

form

buffered aqueous solution

mol wt

~35 kDa

species reactivity

human, mouse, quail, hamster, frog, porcine, rat

concentration

~1.0 mg/mL

technique(s)

ELISA: suitable
dot blot: suitable
immunoblotting: suitable
immunofluorescence: 0.125-0.25 μg/mL using HeLa cells
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SRSF2(6427)

General description

Anti-Splicing Factor SC-35 antibody, Mouse monoclonal (mouse IgG1 isotype) is derived from the SC-35 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from a RBF-DHJ mouse immunized with partially purified spliceosomes prepared from HeLa cells. Splicing Factor SC-35, also known as serine/arginine rich splicing factor 2 (SRSF2) belongs to the family of of serine/arginine-rich (SR) proteins.

Immunogen

partially purified spliceosomes prepared from HeLa cells

Application

Anti-Splicing Factor SC-35 antibody has been used in
  • dot blot
  • enzyme linked immunosorbent assay (ELISA)
  • immunofluorescence
  • immunohistochemistry
  • immunoblotting
  • immunoprecipitation

Biochem/physiol Actions

Splicing Factor SC-35 are critical regulators of constitutive and alternative pre-mRNA splicing. It also controls cellular apoptosis in response to genotoxic stress. Serine/arginine protein kinase 1 (SRPK1) specifically phosphorylates SC-35, resulting in p53 suppression and cyclin D1 upregulation. Elevated levels of SC-35 correlate with aggressive phenotype in several cancer types including lung squamous cell carcinoma and lung adenocarcinoma. SC-35 has oncogenic function in tumor progression of human papilloma virus (HPV)-infected cells.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E2F1 controls alternative splicing pattern of genes involved in apoptosis through upregulation of the splicing factor SC35
Merdzhanova L, et al.
Cell Death and Differentiation, 15(12), 1815-1815 (2008)
Masashi Ueda et al.
Molecular brain, 13(1), 80-80 (2020-05-26)
Zhu-Tokita-Takenouchi-Kim (ZTTK) syndrome, a rare congenital anomaly syndrome characterized by intellectual disability, brain malformation, facial dysmorphism, musculoskeletal abnormalities, and some visceral malformations is caused by de novo heterozygous mutations of the SON gene. The nuclear protein SON is involved in
Carlos Rivera et al.
Epigenetics & chromatin, 14(1), 51-51 (2021-11-26)
Nuclear processes such as transcription and RNA maturation can be impacted by subnuclear compartmentalization in condensates and nuclear bodies. Here, we characterize the nature of nuclear granules formed by REST corepressor 2 (RCOR2), a nuclear protein essential for pluripotency maintenance
Shengyan Gao et al.
Proceedings of the National Academy of Sciences of the United States of America, 119(25), e2206046119-e2206046119 (2022-06-16)
Nuclear speckles are non-membrane-bound organelles known as storage sites for messenger RNA (mRNA) processing and splicing factors. More recently, nuclear speckles have also been implicated in splicing and export of a subset of mRNAs, including the influenza virus M mRNA

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service