Skip to Content
Merck
All Photos(6)

Key Documents

HPA019816

Sigma-Aldrich

Anti-ARHGAP45 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

HA-1, HMHA1, KIAA0223

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Estimated to ship on17 April 2025



Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$1,040.00


Estimated to ship on17 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

QRAEVVVQVPYLEAGEAVVYPLQEAAADGCRESRVVSNDSDSDLEEASELLSSSEASALGHLSFLEQQQSEASLEVASGSHSGSEEQLEATAREDGDGDEDGPAQQLSGFNTNQSNNV

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HMHA1(23526)

General description

HMHA1 (Histocompatibility minor HA-1) is an autosomal minor H antigen consisting of RhoGTPase activating protein (GAP) domain at the C-terminal end and a Bin/Amphiphysin/Rvs (BAR) domain at the N-terminal end. It is located on chromosome 19p13.3. It is mainly expressed in hematopoietic system, comprising normal and leukemic cells, including progenitor cells.

Immunogen

Rho GTPase activating protein 45 recombinant protein epitope signature tag (PrEST)

Application

Anti-HMHA1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

HMHA1 (Histocompatibility (minor) HA-1) acts as a novel RhoGAP factor involved in the regulation of actin cytoskeleton organization. It is associated with several essential functions in normal hematopoietic and cancer cells GTPase activity, cytoskeletal remodelling and cell spreading.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71221

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Le He et al.
EMBO reports, 22(4), e52196-e52196 (2021-03-16)
T and B cells continually recirculate between blood and secondary lymphoid organs. To promote their trans-endothelial migration (TEM), chemokine receptors control the activity of RHO family small GTPases in part via GTPase-activating proteins (GAPs). T and B cells express several
Mohamed Hichem Sellami et al.
Clinics (Sao Paulo, Brazil), 65(11), 1099-1103 (2011-01-19)
Minor histocompatibility antigen HA-1 (MiHAg-HA-1) disparity between a patient and his or her human leukocyte antigen (HLA) genoidentical donor has been widely associated with an increased risk of graft-versus-host disease following allogeneic hematopoietic stem cell transplantation. To examine the effect
Peng Xu et al.
Journal of dermatological science, 88(2), 216-224 (2017-09-25)
Remodeling of cytoskeleton plays an important role in development of multiple cancers, including melanoma. As a group of F-actin regulators, the Ras homology (Rho) GTPase-activating proteins (ARHGAPs) were reported by accumulating studies as a set of significant mediators in cell
Bart-Jan de Kreuk et al.
PloS one, 8(9), e73962-e73962 (2013-10-03)
The human minor Histocompatibility Antigen HMHA-1 is a major target of immune responses after allogeneic stem cell transplantation applied for the treatment of leukemia and solid tumors. The restriction of its expression to hematopoietic cells and many solid tumors raised

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service