Skip to Content
Merck
All Photos(2)

Documents

SAB5600251

Sigma-Aldrich

Anti-N6-Methyladenosine (m6A) antibody, Rabbit monoclonal

recombinant, expressed in HEK 293 cells, clone RM362, purified immunoglobulin

Synonym(s):

m6A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

recombinant

expressed in HEK 293 cells

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RM362, monoclonal
recombinant monoclonal

form

buffered aqueous glycerol solution

species reactivity

vertebrates, human

technique(s)

ELISA: 0.1-3 μg/mL
dot blot: 0.5-2 μg/mL
immunocytochemistry: 0.1— 3 μg/mL
immunoprecipitation (IP): .2-5 μg/mL (Methylated RNA)

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... METTL3(56339)

Specificity

This antibody reacts to N6-Methyladenosine (m6A) in both RNA and DNA. No cross reactivity with non-methylated adenosine.

Immunogen

BSA-conjugated N6-Methyladenosine

Physical form

Solution in phosphate buffered saline containing 50% Glycerol/PBS with 1% BSA and 0.09% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fang Wang et al.
Chinese medicine, 18(1), 122-122 (2023-09-22)
Curdione is a sesquiterpene isolated from Curcumae Rhizoma that possesses high biological activity and extensive pharmacological effects. As a traditional Chinese medicine, Curcumae Rhizoma can inhibit the development of many types of cancer, especially colorectal cancer. However, the anti-colorectal mechanism
Shuai Ren et al.
Annals of translational medicine, 11(1), 15-15 (2023-02-11)
Keloid is a dermal fibrotic disease characterized by excessive proliferation of dermal fibroblasts and deposition of excessive collagen. N6-methyladenosine (m6A) plays a significant role in numerous physiological and pathological regulatory processes in the human body. Fat mass and obesity-associated protein
Yufan Ying et al.
Molecular cancer, 23(1), 79-79 (2024-04-25)
R-loops are prevalent three-stranded nucleic acid structures, comprising a DNA-RNA hybrid and a displaced single-stranded DNA, that frequently form during transcription and may be attributed to genomic stability and gene expression regulation. It was recently discovered that RNA modification contributes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service